missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford AMPSO
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- pKa (25°C): 9.0
- The working pH range in aqueous solution is 8.3 - 9.7
Specifications
Specifications
| Chemical Name or Material | AMPSO |
| CAS | 68399-79-1 |
| Quantity | 100 g |
| Molecular Formula | C7H17NO5S |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction