missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals AMPSO, 0.2M buffer soln., pH 9.0

Product Code. 15454729
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454729 100 mL 100mL
15464729 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454729 Supplier Thermo Scientific Chemicals Supplier No. J62328.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102029-60-7 | C7H16NNaO5S | 249.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102029-60-7
Molecular Formula C7H16NNaO5S
Molecular Weight (g/mol) 249.257
MDL Number MFCD00067500
InChI Key UVGYCHPTWHWEPZ-UHFFFAOYSA-M
PubChem CID 44119996
IUPAC Name sodium;2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonate
SMILES CC(C)(CO)NCC(CS(=O)(=O)[O-])O.[Na+]

Specifications

Color Colorless
Concentration 0.2 M
pH 9.0
Physical Form Liquid
Quantity 100 mL
Odor Odorless
Chemical Name or Material AMPSO buffer soln.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.