missing translation for 'onlineSavingsMsg'
Learn More

Amprolium hydrochloride, MedChemExpress

Product Code. 16458526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458526 Supplier Medchem Express Supplier No. HYB0937A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amprolium hydrochloride is a coccidiostat used in poultry, is a thiamine analogue and blocks the thiamine transporter of Eimeria species by blocking thiamine uptake it prevents carbohydrate synthesis.

Amprolium hydrochloride is a coccidiostat used in poultry, is a thiamine analogue and blocks the thiamine transporter of Eimeria species by blocking thiamine uptake it prevents carbohydrate synthesis.

Specifications

Chemical Name or Material Amprolium hydrochloride
Physical Form Solid
CAS 137-88-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H20Cl2N4
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : ≥ 200 mg/mL (634.44 mM)
SMILES CC1=CC=CC=[N+]1CC2=CN=C(CCC)N=C2N.[Cl-].Cl
Molecular Weight (g/mol) 315.24
Formula Weight 315.24
Percent Purity 99.11%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.