missing translation for 'onlineSavingsMsg'
Learn More

Ampkinone, MedChemExpress

Product Code. 16475378
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475378 10 mg 10mg
16468097 5 mg 5mg
16460176 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16475378 Supplier Medchem Express Supplier No. HY12831
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ampkinone is an indirect AMP-activated protein kinase (AMPK) activator.

Ampkinone is an indirect AMP-activated protein kinase (AMPK) activator.

Specifications

Chemical Name or Material Ampkinone
CAS 1233082-79-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H23NO6
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (98.91 mM; Need ultrasonic)
SMILES O=C1N(C2=CC=C(C(C3=CC=CC=C3)=O)C=C2)C(C4=C1C=CC5=C4C(C)(C)OC6=C(O)C(OC)=CC=C65)=O
Molecular Weight (g/mol) 505.52
Formula Weight 505.52
Percent Purity 99.25%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.