missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMPK gamma-2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517561
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
AMP-activated protein kinase (AMPK) is a heterotrimeric protein composed of a catalytic alpha subunit, a noncatalytic beta subunit, and a noncatalytic regulatory gamma subunit. Various forms of each of these subunits exist, encoded by different genes. AMPK is an important energy-sensing enzyme that monitors cellular energy status and functions by inactivating key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This gene is a member of the AMPK gamma subunit family and encodes a protein with four cystathionine beta-synthase domains. Mutations in this gene have been associated with ventricular pre-excitation (Wolff-Parkinson-White syndrome), progressive conduction system disease and cardiac hypertrophy. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| AMPK gamma-2 | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q91WG5, Q9UGJ0 | |
| PRKAG2 | |
| Synthetic peptide correspoding to residues surrounding Ser60 of human AMPKgamma2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Bovine | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PRKAG2 | |
| 2410051C13Rik; 5'-AMP-activated protein kinase subunit gamma-2; 5-AMP-activated protein kinase, gamma-2 subunit; AAKG; AAKG2; adenosine monophosphate-activated protein kinase gamma 2-subunit; AI854673; AMP-activated protein kinase gamma2 subunit; ampk gamma; AMPK gamma 2; AMPK gamma2; AMPK gamma-2 chain; AMPK subunit gamma-2; CMH6; H91620p; OTTHUMP00000211956; PRKAG2; protein kinase AMP-activated non-catalytic subunit gamma 2; protein kinase, AMP-activated, gamma 2 non-catalytic subunit; WPWS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108099, 373545, 504219, 51422 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction