missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK gamma 1 Rabbit anti-Human, Mouse, Rat, Clone: S08-7D5, Novus Biologicals™
Rabbit Monoclonal Antibody
£451.00
Specifications
| Antigen | AMPK gamma 1 |
|---|---|
| Clone | S08-7D5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
AMPK gamma 1 Monoclonal antibody specifically detects AMPK gamma 1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| AMPK gamma 1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Hypoxia | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 5571 | |
| IgG | |
| Affinity-purified |
| S08-7D5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 5'-AMP-activated protein kinase subunit gamma-1, 5'-AMP-activated protein kinase, gamma-1 subunit, AMPK gamma1, AMPK gamma-1 chain, AMPK subunit gamma-1, AMPKg, MGC8666, protein kinase, AMP-activated, gamma 1 non-catalytic subunit | |
| A synthetic peptide of human AMPK gamma 1 (Uniprot#: P54619) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title