missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK gamma 1 Antibody (5E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26370-100ul
This item is not returnable.
View return policy
Description
AMPK gamma 1 Monoclonal antibody specifically detects AMPK gamma 1 in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| AMPK gamma 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 5E5 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| 5'-AMP-activated protein kinase subunit gamma-1, 5'-AMP-activated protein kinase, gamma-1 subunit, AMPK gamma1, AMPK gamma-1 chain, AMPK subunit gamma-1, AMPKg, MGC8666, protein kinase, AMP-activated, gamma 1 non-catalytic subunit | |
| A synthesized peptide derived from Human AMPK gamma 1 [UniProt P54619] | |
| 100 μL | |
| Cancer, Hypoxia | |
| 5571 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction