missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AMPK beta 1 Monoclonal antibody specifically detects AMPK beta 1 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | AMPK beta 1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 1092531 |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 5'-AMP-activated protein kinase beta-1 subunit, AMP-activated protein kinase beta subunit, AMPK, AMPK beta -1 chain, AMPK beta 1,5'-AMP-activated protein kinase subunit beta-1, AMPK subunit beta-1, AMPKb, HAMPKb, MGC17785, protein kinase, AMP-activated, beta 1 non-catalytic subunit, protein kinase, AMP-activated, noncatalytic, beta-1 |
| Host Species | Mouse |
| Immunogen | E. coli- derived recombinant human AMPKbeta1, Met1-Ile270, Accession # Q9Y478 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?