missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMPK alpha-2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521494
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, NIH-3T3. Predicted reactivity: Mouse (98%), Rat (98%), Pig (99%), Chicken (96%), Sheep (98%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a catalytic subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. Studies of the mouse counterpart suggest that this catalytic subunit may control whole-body insulin sensitivity and is necessary for maintaining myocardial energy homeostasis during ischemia.
Specifications
| AMPK alpha-2 | |
| Polyclonal | |
| Unconjugated | |
| PRKAA2 | |
| 2310008I11Rik; 5'-AMP-activated protein kinase catalytic subunit alpha-2; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; A830082D05; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP-activated protein kinase; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMPK; AMPK alpha 2; AMPK alpha-2 chain; AMPK subunit alpha-2; AMPK2; AMPKa2; AMPKalpha2; AMPK-alpha-2 chain; EC 2.7.11.1; HMGCR kinase; hydroxymethylglutaryl-CoA reductase kinase; PRKAA; Prkaa2; protein kinase AMP-activated catalytic subunit alpha 2; protein kinase, AMP-activated, alpha 2 catalytic subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 108079, 5563, 78975 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.96 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P54646, Q09137, Q8BRK8 | |
| PRKAA2 | |
| Recombinant protein encompassing a sequence within the center region of human AMPK alpha 2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction