missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK alpha 2, p Ser491 Antibody (3H10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26401-50ul
This item is not returnable.
View return policy
Description
AMPK alpha 2 Monoclonal antibody specifically detects AMPK alpha 2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| AMPK alpha 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 3H10 | |
| Western Blot 1:500-1:5000, ELISA | |
| 5'-AMP-activated protein kinase catalytic subunit alpha-2, AMPK subunit alpha-2, AMPK2, AMPK5'-AMP-activated protein kinase, catalytic alpha-2 chain, AMPK-alpha-2 chain, EC 2.7.11, EC 2.7.11.1, PRKAA, protein kinase, AMP-activated, alpha 2 catalytic subunit | |
| A synthesized peptide derived from Human AMPK alpha 2 [p Ser491] [UniProt P54646] | |
| 50 μL | |
| Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Translation Control | |
| 5563 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction