missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK alpha 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£148.00 - £367.00
Specifications
| Antigen | AMPK alpha 1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Knockout Validated |
| Applications | Western Blot, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18310336
|
Novus Biologicals
NBP3-15610-20UL |
20 μg |
£148.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18327972
|
Novus Biologicals
NBP3-15610-100UL |
100 μg |
£367.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AMPK alpha 1 Polyclonal antibody specifically detects AMPK alpha 1 in Human samples. It is validated for Western Blot, KnockDownSpecifications
| AMPK alpha 1 | |
| Western Blot, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Signal Transduction, Translation Control | |
| PBS, 50% glycerol, pH7.3 | |
| 5562 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5'-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5'-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit | |
| Recombinant protein of human AMPK alpha 1. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title