missing translation for 'onlineSavingsMsg'
Learn More

AMPK activator 1, MedChemExpress

Product Code. 16423309
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16423309 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16423309 Supplier Medchem Express Supplier No. HYU00292
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress AMPK activator 1 is an AMPK activator extracted from patent WO2013116491A1, compound No.1-75, has an EC50 of <0.1μM.

AMPK activator 1 is an AMPK activator extracted from patent WO2013116491A1, compound No.1-75, has an EC50 of <0.1μM.

Specifications

Chemical Name or Material AMPK activator 1
CAS 1152423-98-7
Color White
Purity Grade Notes Research
Molecular Formula C32H33F3N4O3
Quantity 1 mg
SMILES O=C(C1=CC2=CC(OC3CCN(C4=CC=C(C(F)(F)F)C=C4)CC3)=CC=C2O1)NC5CCN(CC6=CC=NC=C6)CC5
Molecular Weight (g/mol) 578.62
Formula Weight 578.62
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.