missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AMPK alpha-1/beta-2/gamma-3, GST Tag, His Tag Recombinant Protein

Product Code. p-7181504
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15920273

Brand: Invitrogen™ A30485

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This AMPK (A1/B2/G3) recombinant human protein (full length) was expressed in insect cells. AMPK (A1/B2/G3) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active.

AMPK (AMP-activated protein kinase) serine/threonine kinase is a heterotrimeric complex, which acts as sensor of cellular energy levels. The signaling cascades initiated by activating AMPK are critical to regulation of metabolic events in the liver, skeletal muscle, heart, adipose tissue, and pancreas. This AMPK consists of the alpha1, beta2 and gamma3 subunits. While the purified AMPK (A1/B2/G3) complex can be used in a number of different applications, it has been specifically validated for use with our Z'-LYTE™ assay technology to identify novel kinase inhibitors.
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.1mM EDTA, 0.1mM PMSF, 150mM NaCl, 10mM glutathione, 25% glycerol, 0.25mM DTT and no preservative; pH 7.5
Molecular Weight (g/mol) 90.3-56.6-81.4 kDa
Name Human AMPK alpha-1/beta-2/gamma-3, GST Tag, His Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Common Name AMPK alpha-1/beta-2/gamma-3
KinaseFamily CAMKL Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag, His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.