Learn More
Eurofins DiscoverX AMPK (α2, β1, γ1) Protein, Active
Description
Adenosine 5'-monophosphate (AMP)-activated protein kinase (AMPK) is a metabolic master switch, activated by exercise and muscle contraction. AMPK phosphorylate key target proteins that control glucose uptake, oxidation of fatty acids, cholesterol synthesis, lipogenesis, triglyceride synthesis, adipocyte lipolysis, glycolysis and nitric oxide synthesis.
GenBank NM_006252 / GenBank NM_006253 / GenBank NM_002733
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 8.0, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2m M PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 63 kDa (AMPKα2), 30 kDa (AMPKβ1) and 38 kDa (AMPKγ1) |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His / untagged / untagged |
| Expression System | Recombinant enzyme expressed in E. coli cells |
| Protein Form | Truncated |
| Purity or Quality Grade | Combined ≥70% |
| Protein | AMPK (α2, β1, γ1), active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.