missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX AMPK (α2, β1, γ1) Protein, Active

Product Code. p-7228322
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17340764

Brand: Eurofins DiscoverX 14902

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, recombinant, human AMPK (α2, β1, γ1) holoenzyme, comprised of full length N-terminal 6His-tagged AMPKα2, full length untagged AMPKβ1 and full length untagged AMPKγ1, for use in Kinase Assays.

Adenosine 5'-monophosphate (AMP)-activated protein kinase (AMPK) is a metabolic master switch, activated by exercise and muscle contraction. AMPK phosphorylate key target proteins that control glucose uptake, oxidation of fatty acids, cholesterol synthesis, lipogenesis, triglyceride synthesis, adipocyte lipolysis, glycolysis and nitric oxide synthesis.

GenBank NM_006252 / GenBank NM_006253 / GenBank NM_002733

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 8.0, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2m M PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 63 kDa (AMPKα2), 30 kDa (AMPKβ1) and 38 kDa (AMPKγ1)
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His / untagged / untagged
Expression System Recombinant enzyme expressed in E. coli cells
Protein Form Truncated
Purity or Quality Grade Combined ≥70%
Protein AMPK (α2, β1, γ1), active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.