missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ampicillin Trihydrate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Ampicillin Trihydrate |
| CAS | 7177-48-2 |
| Analyte or Component Names | Ampicillin Trihydrate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H19 N3 O4 S . 3 H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ampicillin Trihydrate, BRL-1341 |
| InChI Formula | InChI=1S/C16H19N3O4S.3H2O/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;;;/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);3*1H2/t9-,10-,11+,14-;;;/m1.../s1 |
| Solution Type | Neat |
| SMILES | O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2[C@H]1C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?