missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin

Product Code. 15403659
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15403659

Brand: Thermo Scientific Alfa Aesar J60977.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ampicillin, CAS # 69-53-4, is structurally related to penicillin and is considered to be a semi-synthetic antibiotic with antibacterial activity. | CAS: 69-53-4 | C16H19N3O4S | 349.405 g/mol

Ampicillin is used as asemi-synthetic antibiotic which has antibacterial activity structurally related to penicillin. It is widely used selection reagent for transformed cells expressing β-lactamase. It is also used to prevent and treat a number of bacterial infections. It may also be used to prevent group B streptococcal infection in newborns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ampicillin is a broad-spectrum, semi-synthetic, β-lactam penicillin with bactericidal activity. It acts against several gram-positive and -negative infections
  • This compound binds to and inactivates penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall. This inactivation interferes with the cross-linkage of peptidoglycan chains essential for bacterial cell wall strength and rigidity. Consequently, there is an interruption in the bacterial cell wall synthesis which results in the weakening of the bacterial cell wall causing cell lysis

Application

  • In laboratory experiments, it is commonly used as a selector reagent for transformed cells expressing β-lactamase. The ampicillin-resistant gene (ampR) catalyzes the hydrolysis of the β-lactam ring of ampicillin and naturally detoxifies the drug
  • Ampicillin is commonly used as a selection marker for E. coli and other bacteria during plasmid DNA isolation, protein expression, and gene cloning
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-53-4
Molecular Formula C16H19N3O4S
Molecular Weight (g/mol) 349.405
MDL Number MFCD00005175
InChI Key AVKUERGKIZMTKX-NJBDSQKTSA-N
PubChem CID 6249
ChEBI CHEBI:28971
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C

Specifications

Type Ampicillin
Physical Form Powder
Melting Point 208°C (decomposition)
Color White
Quantity 25 g
Beilstein 1090925
Merck Index 14,586
Solubility Information In 1 M NH4OH,50mg/ml
Formula Weight 349.4
Chemical Name or Material Ampicillin
How can I prepare LB-ampicillin using Ampicillin (Cat. No. J60977)?

We recommend the following the instructions to prepare LB-ampicillin:

  1. Dissolve the following in 500 mL dH2O:
    • 5g tryptone
    • 2.5g yeast extract
    • 5.0g NaCl
    • 7.5g agar
    • OR
    • 20g pre-mixed LB agar powder
  2. Boil the solution on a hot plate while stirring for 1-2 minutes.
  3. Autoclave the solution for 20 minutes and let cool to 50-60°C.
  4. Add 25 mg ampicillin and swirl or stir on a hot plate to mix.
  5. Pour approximately 10 mL of molten LB agar into each plate.
  6. Allow plates to solidify for approximately 20 minutes.
Can Ampicillin (Cat. No. J60977), be used for administration to or consumption by humans or animals?

Ampicillin, (Cat. No. J60977), is an RUO (Research Use Only) chemical. It is not intended for animal or human diagnostic or therapeutic use.

compliance-icons
Product Identifier
  • Ampicillin
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ampicillin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.