missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin Sodium Salt, TRC

Product Code. 30010251
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010251 50 g 50g
30004344 5 g 5g
17914235 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010251 Supplier Toronto Research Chemicals Supplier No. TRCA63431550G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-52-3
Molecular Formula C16 H18 N3 O4 S . Na
Molecular Weight (g/mol) 371.39
Synonym Ampicillin Sodium, Piperacillin Sodium Imp. A (EP) as Sodium Salt, Piperacillin Imp. A (EP) as Sodium Salt, (2S,5R,6R)-6-[[(2R)-Aminophenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monosodium salt, D-(-)-6-(2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monosodium salt, [2S-[2α,5α,6β(S*)]]-6-[(Aminophenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monosodium salt, Alpen N, Amcill S, Ampi-Dry 5000, Ampicillin sodium, Ampicillin sodium salt, Ampicillin-Na, Ampicin, Anhypen, Binotal sodium, Britapen injection, Cilleral, D-α-Aminobenzylpenicillin sodium salt, Monosodium ampicillin, Omnipen N, Pen A/N, Penbritin S, Pentrex, Polycillin N, Principen N, Sodium 6-(2-amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Sodium 6-[D-α-aminophenylacetamido]penicillanoate, Sodium D-α-aminobenzylpenicillanate, Sodium P-50, Sodium ampicillin, Sodium ampicillinate, Sodium binotal, Solampi
IUPAC Name sodium;(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2[C@H]1C(=O)[O-]

Specifications

Quantity 50 g
InChI Formula InChI=1S/C16H19N3O4S.Na/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);/q;+1/p-1/t9-,10-,11+,14-;/m1./s1
Formula Weight 371.0916
Chemical Name or Material Ampicillin Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.