missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin sodium salt

Product Code. 15475979
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475979 10 g 10g
15465979 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475979 Supplier Thermo Scientific Alfa Aesar Supplier No. J63807.09
  • ONLINE EXCLUSIVE PRICE
    £107.10
    £83.65 / 10g
    Save £23.45
    21% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ampicillin sodium salt, CAS # 69-52-3, is the sodium salt form of ampicillin, a semisynthetic derivative of aminopenicillin compound that shows antibacterial activity against both gram-positive and gram-negative bacteria. | CAS: 69-52-3 | C16H21N3NaO4S | 374.411 g/mol

Ampicillin sodium salt is a reagent for transformed cells expressing beta-lactamase. It acts as a structural analogue of acyl-D-alanyl-D-alanine, acylates the transpeptidase enzyme and prevents the cross-linking of the peptidoglycan of the cell wall necessary for the growth of the bacterium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Ampicillin sodium salt is a semi-synthetic derivative of penicillin with a broad-spectrum antibacterial activity against several species, including Bacillus subtilis, Staphylococcus aureus, Escherichia coli, and Ralstonia solanacearum

• It can inhibit bacterial cell wall synthesis by binding to penicillin-binding proteins, inhibiting peptidoglycan synthesis, which is an essential component of the bacterial cell wall

Application

• Ampicillin sodium salt has been used as a reagent for transformed cells that express β-lactamase

• It can be used to select for ampicillin resistance in mutated and transformed cells

• Ampicillin sodium salt acts as a structural analog of acyl-D-alanyl-D-alanine, acylates the transpeptidase enzyme preventing the cross-linking of the cell wall peptidoglycan, which is necessary for the growth of the bacterium

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-52-3
Molecular Formula C16H21N3NaO4S
Molecular Weight (g/mol) 374.411
MDL Number MFCD00064313
InChI Key BSFVNXCYXDYHOD-ZQDFAFASSA-N
PubChem CID 131673879
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]

Specifications

Type Ampicillin Sodium Salt
Physical Form Powder
Melting Point 215°C
Color White
Quantity 10 g
Beilstein 4119211
Merck Index 14,586
Solubility Information Freely soluble in water; Sparingly soluble in acetone
Formula Weight 371.39
Chemical Name or Material Ampicillin sodium salt
compliance-icons
Product Identifier
  • Ampicillin sodium salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Ampicillin sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.