missing translation for 'onlineSavingsMsg'
Learn More

Melford Ampicillin Sodium Salt

Product Code. 30018037
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
1 kg
Unit Size:
100g
10g
1g
1kg
25g
5g
This item is not returnable. View return policy

Product Code. 30018037

Brand: Melford A40040100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ampicillin, Sodium Salt is an antibiotic that belongs to the penicillin class of antibiotics and is effective against a broad range of Gram-positive and Gram-negative bacteria.

Ampicillin, Sodium Salt is an antibiotic that belongs to the penicillin class of antibiotics and is effective against a broad range of Gram-positive and Gram-negative bacteria. Ampicillin is a semi-synthetic derivative of penicillin and is known for its stability and efficacy.

In the field of molecular biology and genetic engineering, Ampicillin, Sodium Salt is commonly used as a selective agent in plasmid-based cloning experiments. It functions by inhibiting bacterial cell wall synthesis, leading to cell death.

Ampicillin, Sodium Salt is also used in microbiology and cell culture applications. It is effective in preventing the growth of bacterial contaminants in cell cultures, ensuring the purity of cell lines. Additionally, it is employed for the isolation and maintenance of strains with known or engineered resistance to Ampicillin.

One advantage of Ampicillin, Sodium Salt is its stability in aqueous solutions, making it suitable for long-term storage and easy handling. It is available in powdered form and can be easily dissolved in water or other appropriate solvents for use in various applications.

Specifications

Chemical Name or Material Ampicillin, Sodium Salt
CAS 69-52-3
Quantity 100 g
Molecular Formula C16H18N3NaO4S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.