missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin Sodium, Mikromol™

Product Code. 17970366
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970366 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17970366 Supplier Mikromol Supplier No. MM0253.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ampicillin Sodium
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ampicillin Sodium
CAS 69-52-3
Analyte or Component Names Ampicillin Sodium
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H18 N3 O4 S . Na
Certifications/Compliance ISO 17034
Synonym Piperacillin Sodium Imp. A (EP), Piperacillin Imp. A (EP), Ampicillin Sodium, Piperacillin Sodium Imp. A (EP) as Sodium Salt, Piperacillin Imp. A (EP) as Sodium Salt, (2S,5R,6R)-6-[[(2R)-Aminophenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monosodium salt, D-(-)-6-(2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monosodium salt, [2S-[2alpha,5alpha,6beta(S*)]]-6-[(Aminophenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monosodium salt, Alpen N, Amcill S, Ampi-Dry 5000, Ampicillin sodium, Ampicillin sodium salt, Ampicillin-Na, Ampicin, Anhypen, Binotal sodium, Britapen injection, Cilleral, D-alpha-Aminobenzylpenicillin sodium salt, Monosodium ampicillin, Omnipen N, Pen A/N, Penbritin S, Pentrex, Polycillin N, Principen N, Sodium 6-(2-amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Sodium 6-[D-alpha-aminophenylacetamido]penicillanoate, Sodium D-alpha-aminobenzylpenicillanate, Sodium P-50, Sodium ampicillin, Sodium ampicillinate, Sodium binotal, Solampi, Piperacillin Impurity A, Piperacillin Impurity A as Sodium Salt, Piperacillin Sodium Impurity A as Sodium Salt, Sultamicillin Impurity C as Sodium Salt, Sultamicillin Tosilate Dihydrate Impurity B as Sodium Salt, Piperacillin Impurity A as Sodium Salt, Sultamicillin Impurity B as Sodium Salt
InChI Formula InChI=1S/C16H19N3O4S.Na/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);/q;+1/p-1/t9-,10-,11+,14-;/m1./s1
SMILES [Na+].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2[C@H]1C(=O)[O-]
IUPAC Name sodium;(2S,5R,6R)-6-[[(2R)-2-amino-2-phenyl-acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 371.39
Quantity 500 mg
Formula Weight 371.0916 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.