missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin sodium, MedChemExpress

Product Code. 16410978
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16410978

Brand: Medchem Express HYB0522A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ampicillin sodium (D-(-)-α-Aminobenzylpenicillin sodium salt) is a broad-spectrum beta-lactam antibiotic against a variety of gram-positive and gram-negative bacteria.

Ampicillin sodium (D-(-)-α-Aminobenzylpenicillin sodium salt) is a broad-spectrum beta-lactam antibiotic against a variety of gram-positive and gram-negative bacteria.

Specifications

Chemical Name or Material Ampicillin sodium
Physical Form Powder
CAS 69-52-3
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C16H18N3NaO4S
For Use With (Application) COVID-19-immunoregulation
Synonym D-(-)-α-Aminobenzylpenicillin sodium salt
Solubility Information H2O : ≥ 200 mg/mL (538.52 mM) ∣DMSO : 200 mg/mL (538.52 mM; Need ultrasonic)
SMILES O=C([C@@H]1N(C2=O)[C@]([C@@H]2NC([C@@H](C3=CC=CC=C3)N)=O)([H])SC1(C)C)O[Na]
Molecular Weight (g/mol) 371.39
Formula Weight 371.38
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.