missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin sodium, MedChemExpress

Product Code. 16492209
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492209 1 g 1g
16410978 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492209 Supplier Medchem Express Supplier No. HYB0522A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ampicillin sodium (D-(-)-α-Aminobenzylpenicillin sodium salt) is a broad-spectrum beta-lactam antibiotic against a variety of gram-positive and gram-negative bacteria.

Ampicillin sodium (D-(-)-α-Aminobenzylpenicillin sodium salt) is a broad-spectrum beta-lactam antibiotic against a variety of gram-positive and gram-negative bacteria.

Specifications

Chemical Name or Material Ampicillin sodium
Physical Form Powder
CAS 69-52-3
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C16H18N3NaO4S
For Use With (Application) COVID-19-immunoregulation
Synonym D-(-)-α-Aminobenzylpenicillin sodium salt
Solubility Information H2O : ≥ 200 mg/mL (538.52 mM) ∣DMSO : 200 mg/mL (538.52 mM; Need ultrasonic)
SMILES O=C([C@@H]1N(C2=O)[C@]([C@@H]2NC([C@@H](C3=CC=CC=C3)N)=O)([H])SC1(C)C)O[Na]
Molecular Weight (g/mol) 371.39
Formula Weight 371.38
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.