Learn More
Thermo Scientific Chemicals Ampicillin, Ready-to-Use aqueous soln., 100mg/mL, 0.2 micron filtered
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Ampicillin is a broad-spectrum, semi-synthetic, β-lactam penicillin with bactericidal activity. It acts against several gram-positive and -negative infections
- This compound binds to and inactivates penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall. This inactivation interferes with the cross-linkage of peptidoglycan chains essential for bacterial cell wall strength and rigidity. Consequently, there is an interruption in the bacterial cell wall synthesis which results in the weakening of the bacterial cell wall causing cell lysis
Application
- In laboratory experiments, it is commonly used as a selector reagent for transformed cells expressing β-lactamase. The ampicillin-resistant gene (ampR) catalyzes the hydrolysis of the β-lactam ring of ampicillin and naturally detoxifies the drug
- Ampicillin is commonly used as a selection marker for E. coli and other bacteria during plasmid DNA isolation, protein expression, and gene cloning
Specifications
Specifications
| Type | Ampicillin |
| Physical Form | Liquid |
| Odor | Odorless |
| Color | Yellow |
| Quantity | 10 mL |
| Sensitivity | Light sensitive |
| Chemical Name or Material | Ampicillin, eady-to-Use aqueous soln., 100mg/mL, 0.2 micron filtered |
Safety and Handling
- Ampicillin, Ready-to-Use aqueous solution
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-[In case of inadequate ventilation] wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: Sodium [2S-[2.alpha.,5.alpha.,6.beta.(S*)]]-6-(amino phenylacetamido)-3,3-dimethyl-7-oxo-4-thia -1-a
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.