missing translation for 'onlineSavingsMsg'
Learn More

Amphotericin B Methyl Ester 90%, TRC

Product Code. 30138590
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138590 1 g 1g
30012207 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138590 Supplier Toronto Research Chemicals Supplier No. TRCA6342551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36148-89-7
Formula Weight 935.5242
InChI Formula InChI=1S/C49H77NO16/c1-29-19-17-15-13-11-9-7-8-10-12-14-16-18-20-37(65-48-46(59)44(50)45(58)33(5)64-48)26-41-43(47(60)62-6)40(56)28-49(61,66-41)27-36(53)24-39(55)38(54)22-21-34(51)23-35(52)25-42(57)63-32(4)31(3)30(29)2/h7-20,29-41,43-46,48,51-56,58-59,61H,21-28,50H2,1-6H3/b8-7+,11-9+,12-10+,15-13+,16-14+,19-17+,20-18+/t29-,30+,31+,32-,33+,34+,35+,36-,37-,38+,39+,40-,41-,43+,44-,45+,46-,48-,49+/m0/s1
IUPAC Name methyl (1S,3R,4E,6E,8E,10E,12E,14E,16E,18S,19R,20R,21S,25R,27R,30R,31R,33S,35R,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-25,27,30,31,33,35,37-heptahydroxy-18,19,20,21-tetramethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,12,14,16-heptaene-38-carboxylate
Molecular Formula C49 H77 N O16
Molecular Weight (g/mol) 936.13
SMILES COC(=O)[C@@H]1[C@@H](O)C[C@@]2(O)C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](C)[C@@H](C)\C=C\C=C\C=C\C=C\C=C\C=C\C=C\[C@@H](C[C@@H]1O2)O[C@@H]3O[C@H](C)[C@@H](O)[C@H](N)[C@@H]3O
Synonym Amphotericin B Monomethyl Ester;
Chemical Name or Material Amphotericin B Methyl Ester

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.