missing translation for 'onlineSavingsMsg'
Learn More

Amphiphysin/AMPH Antibody [DyLight 550], Novus Biologicals™

Product Code. 30732370 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732370 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732370 Supplier Novus Biologicals Supplier No. AF6527L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Amphiphysin/AMPH Polyclonal antibody specifically detects Amphiphysin/AMPH in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amphiphysin/AMPH
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias AMPH1, amphiphysin, amphiphysin (Stiff-Man syndrome with breast cancer 128kDa autoantigen), amphiphysin (Stiff-Mann syndrome with breast cancer 128kD autoantigen), amphiphysin I
Host Species Goat
Immunogen E. coli-derived recombinant human Amphiphysin/AMPH, Asn204-Gly300, Accession # P49418
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immune System Diseases, Immunology, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 273
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.