missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23997-1-AP-150UL
This item is not returnable.
View return policy
Description
AMPD3 is a member of the AMP deaminase gene family. This protein is a highly regulated enzyme that catalyzes the hydrolytic deamination of adenosine monophosphate to inosine monophosphate, a branch point in the adenylate catabolic pathway. The protein is the erythrocyte (E) isoforms, whereas other family members isoforms predominate in muscle (M) and liver (L) cells. Mutations in this gene lead to the clinically asymptomatic, autosomal recessive condition erythrocyte AMP deaminase deficiency.Specifications
| AMPD3 | |
| Polyclonal | |
| Unconjugated | |
| AMPD3 | |
| AMP deaminase 3, AMP deaminase isoform E, AMPD3, Erythrocyte AMP deaminase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11717, 25095, 272 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08739, O09178, Q01432 | |
| AMPD3 | |
| AMPD3 Fusion Protein Ag21199 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction