missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15710-1-AP-20UL
This item is not returnable.
View return policy
Description
Adenosine monophosphate deaminase-2 (EC 3.5.4.6) catalyzes the deamination of AMP to IMP and plays an important role in the purine nucleotide cycle.Specifications
| AMPD2 | |
| Polyclonal | |
| Unconjugated | |
| AMPD2 | |
| AMP deaminase 2, AMP deaminase isoform L, AMPD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109674, 271, 362015 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q01433, Q02356, Q9DBT5 | |
| Ampd2 | |
| AMPD2 Fusion Protein Ag8335 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction