missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AMPD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13473667
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13473667 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13473667 Supplier Invitrogen™ Supplier No. PA529518

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, U87-MG, SK-N-SH, IMR32, SK-N-AS. Predicted reactivity: Mouse (98%), Rat (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Adenosine monophosphate deaminase-2 (EC 3.5.4.6) catalyzes the deamination of AMP to IMP and plays an important role in the purine nucleotide cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMPD2
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Ampd2
Gene Accession No. Q01433, Q02356
Gene Alias 1200014F01Rik; adenosine monophosphate deaminase 2; adenosine monophosphate deaminase 2 (isoform L); adenosine monophosphate deaminase 2 tv2; AI552571; AMP deaminase 2; AMP deaminase isoform L; AMPD; AMPD2; Ampd-2; PCH9; SPG63
Gene Symbols Ampd2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AMPD2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 271, 362015
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.