missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMPD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555316
This item is not returnable.
View return policy
Description
Immunogen sequence: SETSSTKLSH IDEYISSSPT YQTVPDFQRV QITGDYASGV TVEDFEIVCK GLYRALCIRE KYMQKSFQRF PKTPSKYLRN IDGEAWVANE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
AMP deaminase (AMPD) is an allosteric enzyme involved in the regulation of adenosine metabolism. It catalyzes a central reaction in purine nucleotide biosynthesis where AMP is deaminated to IMP, liberating ammonia. There are three functional isoforms of AMPD. AMPD1 (E.C. No 3.5.4.6.) is the skeletal muscle-specific isoform M located in type II muscle fibers, neuromuscular junctions and in capillaries. This protein is the predominant member of AMPD multi-gene family and is considered as the sensor of the cell's changing energy requirements. AMPD1 deficiency causes irregular muscle metabolism due to lower rate of ATP degradation, phosphocreatine hydrolysis and accumulation of lactic acid. Mutated AMPD1 expression has been detected in neuromuscular disorders, exercise-induced skeletal muscle myopathies and congestive heart failures due to coronary artery diseases.
Specifications
| AMPD1 | |
| Polyclonal | |
| Unconjugated | |
| AMPD1 | |
| adenosine monophosphate deaminase 1; adenosine monophosphate deaminase 1 (isoform M); adenosine monophosphate deaminase-1 (muscle); AI553520; AMP deaminase 1; AMP deaminase isoform M; Ampd; Ampd01; AMPD1; Ampd-1; MAD; MADA; MMDD; myoadenylate deaminase; RATAMPD01; RP5-1000E10.1; skeletal muscle AMPD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P23109 | |
| AMPD1 | |
| Recombinant protein corresponding to Human AMPD1. Recombinant protein control fragment (Product #RP-94087). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction