missing translation for 'onlineSavingsMsg'
Learn More

AMPD1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18431808 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431808 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431808 Supplier Novus Biologicals Supplier No. NBP224509AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AMPD1 Polyclonal antibody specifically detects AMPD1 in Human, Mouse, Rat, Bovine, Canine, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMPD1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias adenosine monophosphate deaminase 1, adenosine monophosphate deaminase 1 (isoform M), adenosine monophosphate deaminase-1 (muscle), AMP deaminase 1, AMP deaminase isoform M, AMPD, EC 3.5.4.6, MAD, MADA, Myoadenylate deaminase, skeletal muscle AMPD
Host Species Rabbit
Immunogen A portion of amino acids 140-190 of human AMPDA1 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 270
Target Species Human, Mouse, Rat, Bovine, Canine, Opossum, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.