missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ AMP PNP

Product Code. 18775936
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18775936 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18775936 Supplier R&D Systems™ Supplier No. 6086/10
  • ONLINE EXCLUSIVE PRICE
    £171.00
    £161.70 / 10mg
    Save £9.30
    5% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Kir6 blocker; non-hydrolyzable AMP analog

AMP PNP is a non-hydrolyzable AMP analog. Kir6 (KATP) channel blocker. Inhibits fast axonal transport and stabilizes the interaction of membranous organelles with microtubules.
TRUSTED_SUSTAINABILITY

Specifications

CAS 72957-42-7
Quantity 10 mg
Solubility Soluble to 50mM in water
Product Line Tocris
Molecular Formula C10H13Li4N6O12P3
Formula Weight 529.93g/mol
Color White
Product Type Inhibitor

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.