missing translation for 'onlineSavingsMsg'
Learn More

AMP, 0.2M buffer soln., pH 9.5, Thermo Scientific™

Product Code. 15466129
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466129 100 mL 100mL
15476129 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466129 Supplier Thermo Scientific Alfa Aesar Supplier No. J63988.AE
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 124-68-5
Color Colorless
Concentration 0.2 M
pH 9.5
Physical Form Liquid
Quantity 100 mL
Molecular Formula C4H11NO
Odor Odorless
MDL Number MFCD00008051
Synonym 2-Amino-2-methyl-1-propanol
InChI Key CBTVGIZVANVGBH-UHFFFAOYSA-N
SMILES CC(C)(N)CO
IUPAC Name 2-amino-2-methylpropan-1-ol
Molecular Weight (g/mol) 89.14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.