missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Amoxycillin M.I.C.Evaluator Strips (M.I.C.E.™)

Determine accurate Minimum Inhibitory Concentration (MIC) values of antibiotics using MIC testing strips.

Specifications

Concentration 256-0.015µg/mL
Description Amoxycillin M.I.C.Evaluator Strips (M.I.C.E.™)
Content And Storage 2°C to 8°C
View More Specs

Description

Description

Effortlessly establish accurate Minimum Inhibitory Concentration (MIC) values manually using Thermo Scientific™ Oxoid™ Amoxycillin M.I.C.Evaluator Strips (M.I.C.E.™).

Conveniently packaged M.I.C.E. strips combine the ease of a diffusion test with the accuracy of an MIC test.

Simple to Perform
  • Distinctive gradient format provides excellent contrast with the agar
  • Increased font size makes reading easier
  • Scale conforms to international standards
  • Strips are tailored to meet clinical requirements while maintaining the accuracy required for research

Easy-to-handle
  • The strip is conveniently presented when the sachet is peeled apart
  • Single-strip packaging eliminates the requirement to maintain and quality control opened strips; open one and use it
  • Stability of the individual M.I.C.E. strips is ensured through the inclusion of a desiccant within each sachet
  • 10 and 50 pack sizes offer users maximum flexibility to put together custom panels for testing
  • Easily stackable boxes simplify storage; all antimicrobials are stored in the fridge (2-8°C)

Not all products are available for sale in all territories. Please inquire.

Remelâ„¢ and Oxoidâ„¢ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications

Specifications

256-0.015µg/mL
Amoxycillin M.I.C.Evaluator Strips (M.I.C.E.™)
2°C to 8°C
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1 Year

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.