missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin trihydrate, MedChemExpress

Product Code. 16468498
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468498 10 g 10g
16462189 1 g 1g
16450948 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16468498 Supplier Medchem Express Supplier No. HYB0467B
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amoxicillin (Amoxycillin) trihydrateis an antibiotic with good oral absorption and broad spectrum antimicrobial activity. Amoxicillin trihydrateis inhibits the biosynthesis of polypeptides in the cell wall, thereby inhibiting cell growth.

Amoxicillin (Amoxycillin) trihydrateis an antibiotic with good oral absorption and broad spectrum antimicrobial activity. Amoxicillin trihydrateis inhibits the biosynthesis of polypeptides in the cell wall, thereby inhibiting cell growth.

Specifications

Chemical Name or Material Amoxicillin trihydrate
Physical Form Solid
CAS 61336-70-7
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C16H25N3O8S
For Use With (Application) COVID-19-immunoregulation
Synonym Amoxycillin trihydrate
Solubility Information DMSO : 10 mg/mL (23.84 mM; Need ultrasonic) ∣H2O : 2 mg/mL (4.77 mM; Need ultrasonic)
SMILES OC([C@@H]1N(C2=O)[C@]([C@@H]2NC([C@@H](C3=CC=C(O)C=C3)N)=O)([H])SC1(C)C)=O.[3H2O]
Molecular Weight (g/mol) 419.45
Formula Weight 419.45
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.