missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress L-ANAP hydrochloride
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Specifications
Specifications
| Quantity | 1 mg |
| Storage Requirements | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Solubility | DMSO : 33.33 mg/mL (107.95 mM; Need ultrasonic) ∣ H2O : 1 mg/mL (3.24 mM; Need ultrasonic) |
| Shipping Condition | Room temperature |
| Molecular Weight (g/mol) | 308.76 g/mol |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction