Learn More
Melford Amoxicillin Trihydrate
Description
Amoxicillin is a broad-spectrum, semi-synthetic penicillin antibiotic that is effective against a wide range of bacteria.
Amoxicillin is a broad-spectrum, semi-synthetic penicillin antibiotic that is effective against a wide range of bacteria. The trihydrate form of amoxicillin is a crystalline powder that is often used in laboratory settings.
Amoxicillin inhibits the growth of bacteria by interfering with their cell wall synthesis. It is a versatile antibiotic with low toxicity to mammalian cells, making it a valuable tool in maintaining sterile conditions in cell cultures and studying bacterial physiology.
Researchers use amoxicillin to selectively culture bacteria or to treat bacterial infections in cell cultures.
Specifications
Specifications
| Chemical Name or Material | Amoxicillin Trihydrate |
| CAS | 61336-70-7 |
| Quantity | 25 g |
| Molecular Formula | C16H19N3O5S - 3H2O |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.