missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amoxicillin sodium, MedChemExpress
Description
Amoxicillin (Amoxycillin) sodium is an antibiotic with good oral absorption and broad spectrum antimicrobial activity. Amoxicillin sodium inhibits the biosynthesis of polypeptides in the cell wall, thereby inhibiting cell growth.
Specifications
Specifications
| Chemical Name or Material | Amoxicillin sodium |
| Physical Form | Powder |
| CAS | 34642-77-8 |
| Color | White |
| Quantity | 10 g |
| Purity Grade Notes | Research |
| Molecular Formula | C16H18N3NaO5S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Amoxycillin sodium |
| Solubility Information | H2O : 100 mg/mL (258.14 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (25.81 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?