missing translation for 'onlineSavingsMsg'
Learn More

Amoxapine, TRC

Product Code. 30135790
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135790 100 mg 100mg
30138413 10 g 10g
17993605 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135790 Supplier Toronto Research Chemicals Supplier No. TRCA634230100MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14028-44-5
Molecular Formula C17 H16 Cl N3 O
Molecular Weight (g/mol) 313.78
Synonym 2-Chloro-11-(1-piperazinyl)dibenz[b,f][1,4]oxazepine, Amoxan, Amoxapine, Asendin, Asendis, CL 67772, Demolox, Moxadil, Dibenz[b,f][1,4]oxazepine, 2-chloro-11-(1-piperazinyl)-
IUPAC Name 8-chloro-6-piperazin-1-ylbenzo[b][1,4]benzoxazepine
SMILES Clc1ccc2Oc3ccccc3N=C(N4CCNCC4)c2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H16ClN3O/c18-12-5-6-15-13(11-12)17(21-9-7-19-8-10-21)20-14-3-1-2-4-16(14)22-15/h1-6,11,19H,7-10H2
Formula Weight 313.0982
Chemical Name or Material Amoxapine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.