missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMOTL2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92875-0.1ml
This item is not returnable.
View return policy
Description
AMOTL2 Polyclonal antibody specifically detects AMOTL2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| AMOTL2 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin | |
| angiomotin like 2, KIAA0989, LCCPangiomotin-like protein 2, Leman coiled-coil protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 638-837 of human AMOTL2 (NP_001265612.1). RQFAMDAAATAAAQRDTTLIRHSPQPSPSSSFNEGLLTGGHRHQEMESRLKVLHAQILEKDAVIKVLQQRSRRDPGKAIQGSLRPAKSVPSVFAAAAAGTQGWQGLSSSERQTADAPARLTTDRAPTEEPVVTAPPAAHAKHGSRDGSTQTEGPPDSTSTCLPPEPDSLLGCSSSQRAASLDSVATSRVQDLSDMVEILI | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51421 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction