missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMOTL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539024
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a peripheral membrane protein that is a component of tight junctions or TJs. TJs form an apical junctional structure and act to control paracellular permeability and maintain cell polarity. This protein is related to angiomotin, an angiostatin binding protein that regulates endothelial cell migration and capillary formation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2014].
Specifications
| AMOTL1 | |
| Polyclonal | |
| Unconjugated | |
| AMOTL1 | |
| AMOTL1 | |
| A synthetic peptide derived from the internal region of human AMOTL1 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8IY63, Q9D4H4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 154810, 75723 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction