missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMOTL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116800
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, MCF7, Mouse heart, Mouse lung, Mouse brain, Rat lung, Rat heart Immunogen sequence: LTSSQLAEEK KEEKTWKGSI GLLLGKEHHE HASAPLLPPP PTSALSSIAS TTAASSAHAK TGSKDSSTQT DKSAELFWPS MASLPSRGRL S.
The protein encoded by this gene is a peripheral membrane protein that is a component of tight junctions or TJs. TJs form an apical junctional structure and act to control paracellular permeability and maintain cell polarity. This protein is related to angiomotin, an angiostatin binding protein that regulates endothelial cell migration and capillary formation. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| AMOTL1 | |
| Polyclonal | |
| Unconjugated | |
| AMOTL1 | |
| 2310010G08Rik; 2310067L22Rik; 4932416D09Rik; Amotl1; angiomotin like 1; angiomotin-like 1; angiomotin-like protein 1; JEAP; junction-enriched and associated protein; junction-enriched and -associated protein; mFLJ00155 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 154810, 315430, 75723 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8IY63, Q9D4H4 | |
| AMOTL1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 810-900 of human AMOTL1 (NP_5708991). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction