missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AMOT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540947
This item is not returnable.
View return policy
Description
Peptide sequence: TANKQLAEKE YEGSEDTRKT ISQLFAKNKE SQREKEKLEA ELATARSTNE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.
AMOT (Angiomotin) belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. AMOT plays a central role in tight junction maintenance via the complex formed with ARHGAP17 - which acts by regulating the uptake of polarity proteins at tight junctions. AMOT is expressed predominantly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels.
Specifications
| AMOT | |
| Polyclonal | |
| Unconjugated | |
| AMOT | |
| AMOT; Angiomotin; angiomotin p130 isoform; angiomotin p80 isoform; CAG-2; D0Kist1; KIAA1071; RGD1564027; si:dkey-13f9.6; Sii6; wu:fj21a07 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 154796 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q4VCS5 | |
| AMOT | |
| Synthetic peptide directed towards the N-terminal of human AMOT (aa 517-566). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction