missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMOT Mouse anti-Human, Porcine, Clone: 3H7B3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60156-1-IG-150UL
This item is not returnable.
View return policy
Description
This antibody only recognizes p130.
This gene belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. The encoded protein is expressed predomitly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| AMOT | |
| Monoclonal | |
| 0.87 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4VCS5 | |
| AMOT | |
| peptide | |
| 150 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3H7B3 | |
| Unconjugated | |
| AMOT | |
| AMOT, angiomotin, KIAA1071 | |
| Mouse | |
| Protein A | |
| RUO | |
| 100158213, 154796 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction