missing translation for 'onlineSavingsMsg'
Learn More

Amorolfine hydrochloride, 98%

Product Code. 15525996 Shop All Thermo Scientific Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
missing translation for 'unitSize'
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15525996 1 g 1g
15545996 250 mg 250mg
15535996 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15525996 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 458750010
  • ONLINE EXCLUSIVE PRICE
    £157.00
    £141.65 / 1g
    Save £15.35
    9% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 78613-38-4 | C21H35NO·HCl | 353.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78613-38-4
Molecular Formula C21H35NO·HCl
Molecular Weight (g/mol) 353.97
InChI Key XZKWIPVTHGWDCF-KUZYQSSXSA-N
Synonym amorolfine hydrochloride, amorolfine hcl, loceryl, curanail, locetar, odenil, amorolfin, pekiron, amorolfine hydrochloride jan
PubChem CID 54259
ChEBI CHEBI:59649
IUPAC Name (2R,6S)-2,6-dimethyl-4-[2-methyl-3-[4-(2-methylbutan-2-yl)phenyl]propyl]morpholine;hydrochloride
SMILES CCC(C)(C)C1=CC=C(C=C1)CC(C)CN2CC(OC(C2)C)C.Cl

Specifications

Melting Point 221.0°C to 223.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: 9g/L. Other solubilities: 28 mg/mL dmso, 71 mg/mL ethanol
Formula Weight 353.97
Percent Purity 98%
Chemical Name or Material Amorolfine hydrochloride
compliance-icons compliance-icons
Product Identifier
  • Amorolfine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.