missing translation for 'onlineSavingsMsg'
Learn More

Amodiaquine Dihydrochloride Dihydrate, Mikromol™

Product Code. 30050505
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050505 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050505 Supplier Mikromol Supplier No. MM0617.00
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Amodiaquine Dihydrochloride Dihydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amodiaquine Dihydrochloride Dihydrate
CAS 6398-98-7
Analyte or Component Names Amodiaquine Dihydrochloride Dihydrate
Shipping Condition Room Temperature
Molecular Formula C20 H22 Cl N3 O . 2 Cl H . 2 H2 O
Certifications/Compliance ISO 17034
Synonym Amodiaquine Dihydrochloride Dihydrate, 4-[(7-Chloro-4-quinolinyl)amino]-2-[(diethylamino)methyl]phenol hydrochloride hydrate (1:2:2), 4-[(7-Chloro-4-quinolinyl)amino]-2-[(diethylamino)methyl]phenol dihydrochloride dihydrate, Amodiaquin dihydrochloride dihydrate, Amodiaquine dihydrochloride dihydrate, 4-[(7-Chloro-4-quinolyl)amino]-alpha-(diethylamino)-o-cresol dihydrochloride dihydrate
InChI Formula InChI=1S/C20H22ClN3O.2ClH.2H2O/c1-3-24(4-2)13-14-11-16(6-8-20(14)25)23-18-9-10-22-19-12-15(21)5-7-17(18)19;;;;/h5-12,25H,3-4,13H2,1-2H3,(H,22,23);2*1H;2*1H2
SMILES O.O.Cl.Cl.CCN(CC)Cc1cc(Nc2ccnc3cc(Cl)ccc23)ccc1O
IUPAC Name 4-[(7-chloroquinolin-4-yl)amino]-2-(diethylaminomethyl)phenol;dihydrate;dihydrochloride
Molecular Weight (g/mol) 464.81
Quantity 500 mg
Formula Weight 463.1196 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.