missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amodiaquine Dihydrochloride Dihydrate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Amodiaquine Dihydrochloride Dihydrate |
| CAS | 6398-98-7 |
| Analyte or Component Names | Amodiaquine Dihydrochloride Dihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H22 Cl N3 O . 2 Cl H . 2 H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Amodiaquine Dihydrochloride Dihydrate, 4-[(7-Chloro-4-quinolinyl)amino]-2-[(diethylamino)methyl]phenol hydrochloride hydrate (1:2:2), 4-[(7-Chloro-4-quinolinyl)amino]-2-[(diethylamino)methyl]phenol dihydrochloride dihydrate, Amodiaquin dihydrochloride dihydrate, Amodiaquine dihydrochloride dihydrate, 4-[(7-Chloro-4-quinolyl)amino]-alpha-(diethylamino)-o-cresol dihydrochloride dihydrate |
| InChI Formula | InChI=1S/C20H22ClN3O.2ClH.2H2O/c1-3-24(4-2)13-14-11-16(6-8-20(14)25)23-18-9-10-22-19-12-15(21)5-7-17(18)19;;;;/h5-12,25H,3-4,13H2,1-2H3,(H,22,23);2*1H;2*1H2 |
| Solution Type | Neat |
| SMILES | O.O.Cl.Cl.CCN(CC)Cc1cc(Nc2ccnc3cc(Cl)ccc23)ccc1O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction