missing translation for 'onlineSavingsMsg'
Learn More

Amodiaquine Antibody (6D10), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18412556 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412556 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412556 Supplier Novus Biologicals Supplier No. NBP105106JF549

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Amodiaquine Monoclonal antibody specifically detects Amodiaquine in Nonspecies specific samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amodiaquine
Applications ELISA
Classification Monoclonal
Clone 6D10
Conjugate Janelia Fluor 549
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Amodiaquin;Amodiaquine USP24;Amodiaquine, ring closed
Host Species Mouse
Immunogen 4-amino-2(diethylaminomethyl)phenol a preparation of a carrier protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.