missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Amnionless Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547903
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 10% cross-reactivity with recombinant human Amnionless is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
The protein encoded by this gene is a type I transmembrane protein. It is thought to modulate bone morphogenetic protein (BMP) receptor function by serving as an accessory or coreceptor, and thus facilitates or hinders BMP binding. It is known that the mouse AMN gene is expressed in the extraembryonic visceral endoderm layer during gastrulation, but it is found to be mutated in amnionless mouse. The encoded protein has sequence similarity to short gastrulation (Sog) and procollagen IIA proteins in Drosophila.
Specifications
| Amnionless | |
| Polyclonal | |
| Unconjugated | |
| Amn | |
| 5033428N14Rik; Amn; amnion associated transmembrane protein; amnionless; amnionless homolog; AV002116; PRO1028; Protein amnionless; Soluble protein amnionless; type I transmembrane protein; UNQ513/PRO1028; visceral endoderm-specific type 1 transmembrane protein | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 93835 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q99JB7 | |
| Amn | |
| CHO-derived recombinant mouse Amnionless Ala20-Gly362. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction