missing translation for 'onlineSavingsMsg'
Learn More

Ammonium vanadium oxide, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog Number p-4781795
Click to view available options
Quantity:
50 g
250 g
7803-55-6
H4NO3V
116.98
MFCD00011430
YBVKNHXQSRDWAA-UHFFFAOYSA-M
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
516859
N.O[V](=O)=O
7803-55-6
H4NO3V
116.98
MFCD00011430
YBVKNHXQSRDWAA-UHFFFAOYSA-M
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
516859
N.O[V](=O)=O

Ammonium vanadium oxide is used as a catalyst in organic synthesis, and as an analytical reagent in laboratories. It is employed in the preparation of vanadate. It acts as a sensor for the determination of N-acetyltransferase activity in humans. Further it finds use in dyes, varnishes, indelible inks, drier for paints, and in photography. With copper carbonate, it has been shown to enhance the catalytic activities. Ammonium metavanadate is an environmental-friendly catalyst to prepare alpha-hydroxyphosphonate derivatives in high yield through the reaction of aryl/heteroaryl aldehydes with triethylphosphonate derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 200°C (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.0% min
Solubility Information Slightly soluble in water. Soluble in dilute ammoniaSlightly soluble in water. Soluble in dilute ammonia and diethanolamine. Insoluble in ether and alcohol.
IUPAC Name azanium;oxido(dioxo)vanadium
Formula Weight 116.98
Grade ACS
Packaging Packing group II
Density 2.33 g/cm3
Chemical Name or Material Ammonium vanadium oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'